Thermo Scientific Chemicals

Glyoxal, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: A16144.AP
500 mL, Each
Thermo Scientific Chemicals

Glyoxal, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: A16144.AP
500 mL, Each
Quantity
Catalog number: A16144.AP
also known as A16144-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-22-2
IUPAC Name
oxaldehyde
Molecular Formula
C2H2O2
InChI Key
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
SMILES
O=CC=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay (unspecified)
≥39.5 to ≤40.5%
Identification (FTIR)
Conforms

Description

Glyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.

Solubility
Miscible with water, methanol and dimethylsulfoxide.

Notes
Moisture and heat sensitive. Incompatible with strong oxidizing agents, strong reducing agent, oxygen, peroxides, amides, amines, hydroxy-containing materials, nitric acid and aldehydes.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text