Thermo Scientific Chemicals

Lithium metaborate, dried, 99%, Thermo Scientific Chemicals

Catalog number: A16145.14
25 g, Each
Thermo Scientific Chemicals

Lithium metaborate, dried, 99%, Thermo Scientific Chemicals

Catalog number: A16145.14
25 g, Each
Quantity
Catalog number: A16145.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-69-5
IUPAC Name
tetraboron heptahydrate dilithium
Molecular Formula
B4H14Li2O7
InChI Key
DPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
Appearance (Color)
White
Form
Powder
Assay (unspecified)
>98.5% to <101.5%
Loss on Drying
<2.5% (200?C, constant. wt)

Description

Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

Solubility
Soluble in water(27.5g/L).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alkali metals, acid anhydrides, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text