Thermo Scientific Chemicals

Propionaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A16146.AP
500 mL, Each
Thermo Scientific Chemicals

Propionaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A16146.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16146.AP
also known as A16146-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-38-6
IUPAC Name
propanal
Molecular Formula
C3H6O
InChI Key
NBBJYMSMWIIQGU-UHFFFAOYSA-N
SMILES
CCC=O
Comment
Contains varied amounts of cyclic trimer
Refractive Index
1.3605-1.3655 @ 20?C
Assay (GC)
≥96.0% (sum of monomer and trimer)
Appearance (Color)
Clear colorless
Free acid (titration)
≤1.0%

Description

Propionaldehyde applications include as an intermediate, in pharmaceuticals, perfumes, plastics and polyols or polyhydric alcohols. It is also used as a precursor to trimethylolethane (CH3C(CH2OH)3) through a condensation reaction with formaldehyde, this triol is an important intermediate in the production of alkyd resins. Other applications include reduction to propanol and oxidation to propionic acid. Also used in PEGylated protein purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionaldehyde applications include as an intermediate, in pharmaceuticals, perfumes, plastics and polyols or polyhydric alcohols. It is also used as a precursor to trimethylolethane (CH3C(CH2OH)3) through a condensation reaction with formaldehyde, this triol is an important intermediate in the production of alkyd resins. Other applications include reduction to propanol and oxidation to propionic acid. Also used in PEGylated protein purification.

Solubility
Fully miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text