Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only
General References:
- N.T. Abdel-Ghani,; M.S. Rizka,; M. Mostaf. Extractive determination of ephedrine hydrochloride and bromhexine hydrochloride in pure solutions, pharmaceutical dosage form and urine samples . Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2013, 111131-141.
- Tsutsumi,; Tomoko Watoh,; Koji Kumamoto,; Hiyoshizo Kotsuki,; Tatsuomi Matsuoka. Effects of porphyrins on encystment and excystment in ciliated protozoan Colpoda sp. . Jpn. J. Protozool. V. 2004, 37(2).