Thermo Scientific Chemicals

Iodine monobromide, 98%, Thermo Scientific Chemicals

Catalog number: A16158.22
100 g, Each
Thermo Scientific Chemicals

Iodine monobromide, 98%, Thermo Scientific Chemicals

Catalog number: A16158.22
100 g, Each
Quantity
Catalog number: A16158.22
also known as A16158-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-33-5
IUPAC Name
iodobromane
Molecular Formula
BrI
InChI Key
CBEQRNSPHCCXSH-UHFFFAOYSA-N
SMILES
BrI
Appearance (Color)
Black or grey or purple-blue
Form
Fused or crystalline solid
Assay (Iodometric Titration)
≥97.5 to ≤102.5% (UK-sourced material)
Assay from Supplier's CofA
≥97.5% (US-sourced material)
Comment
Material sourced in UK and US

Description

Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

Solubility
Soluble in water, alcohol, ether, carbon disulfide and glacial acetic acid.

Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, phosphorus, sodium and sodiumoxides and potassium.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text