Thermo Scientific Chemicals

Potassium iodate, 98%, Thermo Scientific Chemicals

Catalog number: A16162.22
100 g, Each
Thermo Scientific Chemicals

Potassium iodate, 98%, Thermo Scientific Chemicals

Catalog number: A16162.22
100 g, Each
Quantity
Catalog number: A16162.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-05-6
IUPAC Name
potassium iodate
Molecular Formula
IKO3
InChI Key
JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES
[K+].[O-][I](=O)=O
Assay (Iodometric Titration)
≥97.5 to ≤102.5%
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Loss on Drying
<1.0% (200?C, constant weight)

Description

Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

Solubility
Soluble in water, potassium iodide solution. Insoluble in alcohol, liquid ammonia and nitric acid.

Notes
Incompatible with aluminum, strong oxidizing agents, sulfides, peroxides, metals and reducing agents. Avoid heat, shock and friction.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text