Thermo Scientific Chemicals

Formaldehyde, 37% w/w aq. soln., stab. with 7-8% methanol

Catalog number: A16163.0F
2500 mL, Each
Thermo Scientific Chemicals

Formaldehyde, 37% w/w aq. soln., stab. with 7-8% methanol

Catalog number: A16163.0F
2500 mL, Each
Quantity
Catalog number: A16163.0F
also known as A16163-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-00-0
IUPAC Name
formaldehyde
Molecular Formula
CH2O
InChI Key
WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES
C=O
Comment
Methanol content 7-8%
Form
Liquid
Appearance (Color)
Clear to slightly turbid colorless
Assay (Aqueous acid-base Titration)
35.5-38.0%
Comment
May become turbid on storage due to slight polymerization

Description

Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and used to make components for the transmission, electrical system, engine block, door panels, axles and brake shoes. It serves as a disinfectant. It also plays an ingredient in glues and as a preservative in medical laboratories as embalming fluid and as a sterilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and used to make components for the transmission, electrical system, engine block, door panels, axles and brake shoes. It serves as a disinfectant. It also plays an ingredient in glues and as a preservative in medical laboratories as embalming fluid and as a sterilizer.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text