Thermo Scientific Chemicals

DL-Histidine monohydrochloride monohydrate, 99%

Catalog number: A16165.06
5 g, Each
Thermo Scientific Chemicals

DL-Histidine monohydrochloride monohydrate, 99%

Catalog number: A16165.06
5 g, Each
Quantity
Catalog number: A16165.06
also known as A16165-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123333-71-1
IUPAC Name
hydrogen 2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrate chloride
Molecular Formula
C6H12ClN3O3
InChI Key
CMXXUDSWGMGYLZ-UHFFFAOYNA-N
SMILES
[H+].O.[Cl-].NC(CC1=CN=CN1)C(O)=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)
7.00-10.14% (0.8-1.2 waters, Theory: 8.59%)

Description

DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text