Thermo Scientific Chemicals

1-Iododecane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A16173.14
25 g, Each
Thermo Scientific Chemicals

1-Iododecane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A16173.14
25 g, Each
Quantity
Catalog number: A16173.14
also known as A16173-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2050-77-3
IUPAC Name
1-iododecane
Molecular Formula
C10H21I
InChI Key
SKIDNYUZJPMKFC-UHFFFAOYSA-N
SMILES
CCCCCCCCCCI
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.4830-1.4870 @ 20?C
Identification (FTIR)
Conforms
Stabilizer
Copper Wire

Description

1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases. Keep the container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text