Thermo Scientific Chemicals

Methyl 3-bromobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A16174.30
250 g, Each
Thermo Scientific Chemicals

Methyl 3-bromobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A16174.30
250 g, Each
Quantity
Catalog number: A16174.30
also known as A16174-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
618-89-3
IUPAC Name
methyl 3-bromobenzoate
Molecular Formula
C8H7BrO2
InChI Key
KMFJVYMFCAIRAN-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(Br)=C1
Appearance (Color)
White to cream
Melting Point (clear melt)
28.0-35.0?C
Form
Fused solid
Assay (GC)
≥98.0%

Description

Methyl 3-bromobenzoate is used as a reagent in organic synthesis. It is used in the synthesis of tetrahydro dibenzazocine, which acts as an inhibitor of 17beta-hydroxysteroid dehydrogenase type 3. Further, it is used to prepare 3-tert-butylphosphanyl-benzoic acid methyl ester by reacting with tert-butyltrimethylsilylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromobenzoate is used as a reagent in organic synthesis. It is used in the synthesis of tetrahydro dibenzazocine, which acts as an inhibitor of 17beta-hydroxysteroid dehydrogenase type 3. Further, it is used to prepare 3-tert-butylphosphanyl-benzoic acid methyl ester by reacting with tert-butyltrimethylsilylphosphine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text