Thermo Scientific Chemicals

n-Nonane, 99%, Thermo Scientific Chemicals

Catalog number: A16177.AE
100 mL, Each
Thermo Scientific Chemicals

n-Nonane, 99%, Thermo Scientific Chemicals

Catalog number: A16177.AE
100 mL, Each
Quantity
Catalog number: A16177.AE
also known as A16177-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-84-2
IUPAC Name
nonane
Molecular Formula
C9H20
InChI Key
BKIMMITUMNQMOS-UHFFFAOYSA-N
SMILES
CCCCCCCCC
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.4040-1.4070 @ 20?C
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.

Solubility
Miscible with ethanol, ether, acetone, benzene, chloroform and hydrogen peroxide. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text