Thermo Scientific Chemicals

Benzyl salicylate, 99%, Thermo Scientific Chemicals

Catalog number: A16184.36
500 g, Each
Thermo Scientific Chemicals

Benzyl salicylate, 99%, Thermo Scientific Chemicals

Catalog number: A16184.36
500 g, Each
Quantity
Catalog number: A16184.36
also known as A16184-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-58-1
IUPAC Name
benzyl 2-hydroxybenzoate
Molecular Formula
C14H12O3
InChI Key
ZCTQGTTXIYCGGC-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1C(=O)OCC1=CC=CC=C1
Form
Liquid as melt
Appearance (Color)
Clear colorless to yellow or pale orange
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Refractive Index
1.5780-1.5820 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text