Thermo Scientific Chemicals

Zinc oxide, heavy, 98%, Thermo Scientific Chemicals

Catalog number: A16188.0B
1000 g, Each
Thermo Scientific Chemicals

Zinc oxide, heavy, 98%, Thermo Scientific Chemicals

Catalog number: A16188.0B
1000 g, Each
Quantity
Catalog number: A16188.0B
also known as A16188-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-13-2
IUPAC Name
oxozinc
Molecular Formula
OZn
InChI Key
XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES
O=[Zn]
Appearance (Color)
White
Complexometric Titration
≥97.5 to ≤102.5%
Form
Powder

Description

Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong alkaline earth metal. Alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text