Thermo Scientific Chemicals

Formaldehyde sodium bisulfite, 95%, Thermo Scientific Chemicals

Catalog number: A16194.22
100 g, Each
Thermo Scientific Chemicals

Formaldehyde sodium bisulfite, 95%, Thermo Scientific Chemicals

Catalog number: A16194.22
100 g, Each
Quantity
Catalog number: A16194.22
also known as A16194-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
870-72-4
IUPAC Name
sodium hydroxymethanesulfonate
Molecular Formula
CH3NaO4S
InChI Key
UOULCEYHQNCFFH-UHFFFAOYSA-M
SMILES
[Na+].OCS([O-])(=O)=O
Form
Powder
Assay (Titration)
≥94.0 to ≤106.0%
Appearance (Color)
White to pale cream

Description

Formaldehyde sodium bisulfite, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It also acts as an pharmaceutical intermediate. It can effectively complex metal ion such as Cu, Zn, Pb, co-depositing with nickel ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formaldehyde sodium bisulfite, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It also acts as an pharmaceutical intermediate. It can effectively complex metal ion such as Cu, Zn, Pb, co-depositing with nickel ion.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic, Keep away from acids Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text