Sodium pyrophosphate decahydrate is used as a buffering agent, an emulsifier, a thickening agent, a food additive and a dispersing agent. It acts as a tartar control agent in tooth paste and dental floss and is used to remove calcium and magnesium from saliva, thereby avoiding its deposition on teeth. It is utilized in detergents to prevent deposition on cloth. It is useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. In antimicrobial studies, it is used as a chelating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium pyrophosphate decahydrate is used as a buffering agent, an emulsifier, a thickening agent, a food additive and a dispersing agent. It acts as a tartar control agent in tooth paste and dental floss and is used to remove calcium and magnesium from saliva, thereby avoiding its deposition on teeth. It is utilized in detergents to prevent deposition on cloth. It is useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. In antimicrobial studies, it is used as a chelating agent.
Solubility
Soluble in water. Insoluble in alcohol and ammonia.
Notes
Incompatible with strong oxidizing agents
RUO – Research Use Only
Zavadovsky, K. V.; Krivonogov, N. G.; Lishmanov, Y. B. Use of Technetium-99m-Labelled Sodium Diphosphate Decahydrate to Assess Right Ventricle Dysfunction in Patients with Pulmonary Embolism. Adv. Mat. Res. 2015, 1084, 536-539.
Yoon, J.; El Mohtar, C. S. Constitutive model parameters of concentrated bentonite suspensions modified with sodium pyrophosphate. J. Mater. Sci. 2015, 50 (15), 5253-5261.