Thermo Scientific Chemicals

Cobalt(II) sulfate heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A16201.22
100 g, Each
Thermo Scientific Chemicals

Cobalt(II) sulfate heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A16201.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16201.22
also known as A16201-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-24-1
IUPAC Name
λ²-cobalt(2+) heptahydrate sulfate
Molecular Formula
CoH14O11S
InChI Key
MEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
Appearance (Color)
Red to dark red or red-brown
Form
Crystals or crystalline powder
Assay from Supplier's CofA
≥97.5 to ≤102.5% (based on Cobalt EA)
Elemental Analysis
Cobalt (Co) : 20.12-21.49% (as heptahydrate)

Description

Cobalt(II) sulfate heptahydrate is used in the preparation of pigments, as well as in the manufacture of other cobalt salts, which is used in porcelains and glass. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) sulfate heptahydrate is used in the preparation of pigments, as well as in the manufacture of other cobalt salts, which is used in porcelains and glass. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text