Tin(II) chloride, anhydrous, 98%
Tin(II) chloride, anhydrous, 98%
Tin(II) chloride, anhydrous, 98%
Tin(II) chloride, anhydrous, 98%
Thermo Scientific Chemicals

Tin(II) chloride, anhydrous, 98%

CAS: 7772-99-8 | Cl2Sn | 189.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A16202.22
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS7772-99-8
IUPAC Nameλ²-tin(2+) dichloride
Molecular FormulaCl2Sn
InChI KeyAXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)White
Ask our AI about this Product
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

Solubility
Soluble in ethanol, acetone, ether and tetrahydrofuran. Insoluble in xylene.

Notes
Tin(II) chloride is very freely soluble in cold water. If the volume of water increases or at hot condition it hydrolyses to form an insoluble hydroxide derivative.
RUO – Research Use Only

General References:

  1. For applications see under dihydrate.
  2. Karami, B.; Eskandari, K.; Zare, Z.; Gholipour, S. A New Access to 1,8-Dioxooctahydroxanthenes Using Yttrium(III) Nitrate Hexahydrate and Tin(II) Chloride Dihydrate as Effective and Reusable Catalysts. Chem. Heterocycl. Compd. 2014, 49 (12), 1715-1722.
  3. Kamali, A. R. Thermokinetic characterisation of tin(II) chloride. J. Therm. Anal. Calorim. 2014, 118 (1), 99-104.
  4. Chia, S. -P.; Li, Y.; Ganguly, R.; So, C. -W. Heteroleptic Germanium(II) and Tin(II) Chlorides Supported by Anionic Ligands Derived from 2,3-Dimethyl-1,4-diaza-1,3-butadiene. Eur. J. Inorg. Chem. 2014, 526-532.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
10253642Certificate of AnalysisJan 13, 2025A16202.22, A16202.36, A16202.0E
10253451Certificate of AnalysisDec 16, 2024A16202.22, A16202.36, A16202.0E
10253450Certificate of AnalysisDec 16, 2024A16202.22, A16202.36, A16202.0E
10249907Certificate of AnalysisMar 08, 2024A16202.22, A16202.36, A16202.0E
10246752Certificate of AnalysisMay 25, 2023A16202.0E
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection