4'-Fluoropropiophenone, 97+%, Thermo Scientific Chemicals
4'-Fluoropropiophenone, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Fluoropropiophenone, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A16219.09
also known as A16219-09
Price (USD)/ Each
31.20
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
31.20
Add to cart
4'-Fluoropropiophenone, 97+%, Thermo Scientific Chemicals
Catalog numberA16219.09
Price (USD)/ Each
31.20
-
Add to cart
Chemical Identifiers
CAS456-03-1
IUPAC Name1-(4-fluorophenyl)propan-1-one
Molecular FormulaC9H9FO
InChI KeyQIJNVLLXIIPXQT-UHFFFAOYSA-N
SMILESCCC(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.0%
Refractive Index1.5025-1.5075 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only