3-Acetylindole, 98%, Thermo Scientific Chemicals
3-Acetylindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Acetylindole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A16221.14
also known as A16221-14
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
3-Acetylindole, 98%, Thermo Scientific Chemicals
Catalog numberA16221.14
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS703-80-0
IUPAC Name1-(1H-indol-3-yl)ethan-1-one
Molecular FormulaC10H9NO
InChI KeyVUIMBZIZZFSQEE-UHFFFAOYSA-N
SMILESCC(=O)C1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Melting Point (clear melt)185.0-194.0?C
Appearance (Color)Pale cream to cream or pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only