Thermo Scientific Chemicals

Benzo-18-crown-6, 97%, Thermo Scientific Chemicals

Catalog number: A16223.03
1 g, Each
Thermo Scientific Chemicals

Benzo-18-crown-6, 97%, Thermo Scientific Chemicals

Catalog number: A16223.03
1 g, Each
Quantity
Catalog number: A16223.03
also known as A16223-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14098-24-9
IUPAC Name
2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Molecular Formula
C16H24O6
InChI Key
DSFHXKRFDFROER-UHFFFAOYSA-N
SMILES
C1COCCOCCOC2=CC=CC=C2OCCOCCO1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
39.0-48.0?C
Appearance (Color)
White to pale yellow
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text