Thermo Scientific Chemicals

N-Boc-L-serine, 98% (dry wt.), may cont. up to 10% water, Thermo Scientific Chemicals

Catalog number: A16224.14
25 g, Each
Thermo Scientific Chemicals

N-Boc-L-serine, 98% (dry wt.), may cont. up to 10% water, Thermo Scientific Chemicals

Catalog number: A16224.14
25 g, Each
Quantity
Catalog number: A16224.14
also known as A16224-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3262-72-4
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
Molecular Formula
C8H15NO5
InChI Key
FHOAKXBXYSJBGX-MLHKIVSYNA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (dry wt. basis)
Form
Powder or crystalline powder
Water Content (Karl Fischer Titration)
<10%
Optical Rotation
-3.5 ? 1? (c=2 in acetic acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text