Thermo Scientific Chemicals

Sodium acetate trihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A16230.0C
10,000 g, Each
Thermo Scientific Chemicals

Sodium acetate trihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A16230.0C
10,000 g, Each
Quantity
Catalog number: A16230.0C
also known as A16230-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6131-90-4
IUPAC Name
sodium acetate trihydrate
Molecular Formula
C2H9NaO5
InChI Key
AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES
O.O.O.[Na+].CC([O-])=O
Comment
May contain small amounts of dark particulate matter from the manufacturing process
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

Sodium acetate trihydrate is used in various applications. It is used as a buffer of pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid in textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium acetate trihydrate is used in various applications. It is used as a buffer of pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid in textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.

Solubility
Soluble in water and alcohol.

Notes
-3H{2}O @ 120°. Incompatible with strong oxidizing agents and halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text