Thermo Scientific Chemicals

4-(Methylamino)phenol sulfate, 99%, Thermo Scientific Chemicals

Catalog number: A16236.0B
1000 g, Each
Thermo Scientific Chemicals

4-(Methylamino)phenol sulfate, 99%, Thermo Scientific Chemicals

Catalog number: A16236.0B
1000 g, Each
Quantity
Catalog number: A16236.0B
also known as A16236-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55-55-0
IUPAC Name
bis(4-(methylamino)phenol); sulfuric acid
Molecular Formula
C14H20N2O6S
InChI Key
ZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
Appearance (Color)
White to pale cream or pale grey
Assay (Aqueous acid-base Titration)
≥98.5% to ≤101.5%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

4-(Methylamino)phenol sulfate, is a is a popular photographic developer used in black & white photography. It is also an excellent developing agent for most continuous tone developer applications, and it has been widely used in published developer formulas as well as commercial products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)phenol sulfate, is a is a popular photographic developer used in black & white photography. It is also an excellent developing agent for most continuous tone developer applications, and it has been widely used in published developer formulas as well as commercial products.

Solubility
It is soluble in water (50 mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text