Polyethylene glycol 1,500, Thermo Scientific Chemicals
Polyethylene glycol 1,500, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylene glycol 1,500, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number A16241.0B
also known as A16241-0B
Price (USD)/ Each
80.65
Online exclusive
89.80 
Save 9.15 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
80.65
Online exclusive
89.80 
Save 9.15 (10%)
Add to cart
Polyethylene glycol 1,500, Thermo Scientific Chemicals
Catalog numberA16241.0B
Price (USD)/ Each
80.65
Online exclusive
89.80 
Save 9.15 (10%)
-
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)≤1.0%
FormWaxy flakes
Identification (FTIR)Conforms
Hydroxyl Value70-80 mg KOH/g
Cell fusion induced by polyethylene glycol(PEG) has become a standard method in somatic cell genetics. PEG promoted cell fusion is involved in the production of hybridoma cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only