Thermo Scientific Chemicals

Polyethylene glycol 1,500, Thermo Scientific Chemicals

Catalog number: A16241.30
250 g, Each
Thermo Scientific Chemicals

Polyethylene glycol 1,500, Thermo Scientific Chemicals

Catalog number: A16241.30
250 g, Each
Quantity
Catalog number: A16241.30
also known as A16241-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Appearance (Color)
White
Water Content (Karl Fischer Titration)
≤1.0%
Form
Waxy flakes
Identification (FTIR)
Conforms
Hydroxyl Value
70-80 mg KOH/g

Description

Cell fusion induced by polyethylene glycol(PEG) has become a standard method in somatic cell genetics. PEG promoted cell fusion is involved in the production of hybridoma cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text