Thermo Scientific Chemicals

Magnesium stearate, Thermo Scientific Chemicals

Catalog number: A16244.30
250 g, Each
Thermo Scientific Chemicals

Magnesium stearate, Thermo Scientific Chemicals

Catalog number: A16244.30
250 g, Each
Quantity
Catalog number: A16244.30
also known as A16244-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-04-0
IUPAC Name
magnesium(2+) dioctadecanoate
Molecular Formula
C36H70MgO4
InChI Key
HQKMJHAJHXVSDF-UHFFFAOYSA-L
SMILES
[Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Form
Powder
Assay from Suppliers CofA
Stearic acid: ≥40.0% (GC)
Impurity content
Lead, Pb: ≤10ppm
Impurity content
Sulfate, SO4: ≤1%
Assay from Suppliers CofA
Stearic & Palmitic acid: ≥90.0% (GC)

Description

Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ing
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text