Thermo Scientific Chemicals

Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: A16247.0F
2500 mL, Each
Thermo Scientific Chemicals

Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: A16247.0F
2500 mL, Each
Quantity
Catalog number: A16247.0F
also known as A16247-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-05-4
IUPAC Name
ethenyl acetate
Molecular Formula
C4H6O2
InChI Key
XTXRWKRVRITETP-UHFFFAOYSA-N
SMILES
CC(=O)OC=C
Form
Liquid
Refractive Index
1.3940-1.3970 @ 20?C
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms

Description

Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text