Thermo Scientific Chemicals

L-Lysine monohydrochloride, 99+%

Catalog number: A16249.18
50 g, Each
Thermo Scientific Chemicals

L-Lysine monohydrochloride, 99+%

Catalog number: A16249.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16249.18
also known as A16249-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
657-27-2
IUPAC Name
(2S)-2,6-diaminohexanoic acid hydrochloride
Molecular Formula
C6H15ClN2O2
InChI Key
BVHLGVCQOALMSV-JEDNCBNOSA-N
SMILES
Cl.NCCCC[C@H](N)C(O)=O
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay (unspecified)
≥99.0 to ≤101.0%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)

Description

L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine.  L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine.  L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text