3-(Trifluoromethyl)benzyl chloride, 98%, Thermo Scientific Chemicals
3-(Trifluoromethyl)benzyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16253.06
also known as A16253-06
Price (USD)/ Each
36.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.40
Add to cart
3-(Trifluoromethyl)benzyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA16253.06
Price (USD)/ Each
36.40
-
Add to cart
Chemical Identifiers
CAS705-29-3
IUPAC Name1-(chloromethyl)-3-(trifluoromethyl)benzene
Molecular FormulaC8H6ClF3
InChI KeyXGASTRVQNVVYIZ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(CCl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4625-1.4665 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only