Thermo Scientific Chemicals

1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A16254.06
5 g, Each
Thermo Scientific Chemicals

1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A16254.06
5 g, Each
Quantity
Catalog number: A16254.06
also known as A16254-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18851-33-7
IUPAC Name
1,10-phenanthroline hydrate hydrochloride
Molecular Formula
C12H11ClN2O
InChI Key
NDLHUHRGAIHALB-UHFFFAOYSA-N
SMILES
O.Cl.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Form
Crystalline powder
Assay from Supplier's CofA
≥98.5% (US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Appearance (Color)
White to pale yellow
Assay (Titration ex Chloride)
≥98.5 to ≤101.5% (UK-sourced material)

Description

1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a tightly closed container in a cool, well-ventilated area away from air and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text