Thermo Scientific Chemicals

Iron(III) hydroxide, alpha-phase, 99+%, Thermo Scientific Chemicals

Catalog number: A16267.30
250 g, Each
Thermo Scientific Chemicals

Iron(III) hydroxide, alpha-phase, 99+%, Thermo Scientific Chemicals

Catalog number: A16267.30
250 g, Each
Quantity
Catalog number: A16267.30
also known as A16267-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20344-49-4
IUPAC Name
ironoylol
Molecular Formula
FeHO2
InChI Key
AEIXRCIKZIZYPM-UHFFFAOYSA-M
SMILES
O[Fe]=O
Appearance (Color)
Yellow to brown
Form
Powder
Assay from Suppliers CofA
Iron analysis: ≥62.2%

Description

Iron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text