Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals
Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number A16269.0B
also known as A16269-0B
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
Add to cart
Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals
Catalog numberA16269.0B
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS28917-82-0
IUPAC Namecalcium 3-(phosphonatooxy)propane-1,2-diol
Molecular FormulaC3H7CaO6P
InChI KeyIWIRHXNCFWGFJE-UHFFFAOYNA-L
SMILES[Ca++].OCC(O)COP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Loss on Drying≤17.65% (150?C/constant weight)
Appearance (Color)White
Complexometric Titration≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only