Thermo Scientific Chemicals

Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A16269.30
250 g, Each
Thermo Scientific Chemicals

Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A16269.30
250 g, Each
Quantity
Catalog number: A16269.30
also known as A16269-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28917-82-0
IUPAC Name
calcium 3-(phosphonatooxy)propane-1,2-diol
Molecular Formula
C3H7CaO6P
InChI Key
IWIRHXNCFWGFJE-UHFFFAOYNA-L
SMILES
[Ca++].OCC(O)COP([O-])([O-])=O
Form
Powder
Loss on Drying
≤17.65% (150?C/constant weight)
Appearance (Color)
White
Complexometric Titration
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text