Thermo Scientific Chemicals

2-(Diethylamino)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: A16274.AU
1000 mL, Each
Thermo Scientific Chemicals

2-(Diethylamino)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: A16274.AU
1000 mL, Each
Quantity
Catalog number: A16274.AU
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-37-8
IUPAC Name
2-(diethylamino)ethan-1-ol
Molecular Formula
C6H15NO
InChI Key
BFSVOASYOCHEOV-UHFFFAOYSA-N
SMILES
CCN(CC)CCO
Appearance (Color)
Clear, colorless to pale yellow
Form
Liquid
Assay from Supplier's CofA
≥98.5% (GC)

Description

Diethylethanolamine can be used as a precursor chemical to procaine. It is used as a corrosion inhibitor in steam and condensate lines by neutralizing carbonic acid and scavenging oxygen. it is used for the synthesis of drugs in the pharmaceutical industry and as a catalyst for the synthesis of polymers in the chemical industry. It is also used as a pH stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylethanolamine can be used as a precursor chemical to procaine. It is used as a corrosion inhibitor in steam and condensate lines by neutralizing carbonic acid and scavenging oxygen. it is used for the synthesis of drugs in the pharmaceutical industry and as a catalyst for the synthesis of polymers in the chemical industry. It is also used as a pH stabilizer.

Solubility
It is miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic , Avoid moisture. Keep away from strong oxidizing agents, strong acids, Copper, Zinc, Iron, Do not store near acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text