Thermo Scientific Chemicals

Hydrocortisone, 98%, Thermo Scientific Chemicals

Catalog number: A16292.03
1 g, Each
Thermo Scientific Chemicals

Hydrocortisone, 98%, Thermo Scientific Chemicals

Catalog number: A16292.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16292.03
also known as A16292-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-23-7
IUPAC Name
(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular Formula
C21H30O5
InChI Key
JYGXADMDTFJGBT-VWUMJDOOSA-N
SMILES
C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Form
Powder
Comment
Material sourced in the U.S. and in other countries
Appearance (Color)
White
Assay (HPLC)
≥97.5%
Optical Rotation
153.0 ± 3.5° (C=1 in dioxane, non-U.S. sourced material)

Description

Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

Solubility
It is soluble in DMSO and ethanol.

Notes
Protect against light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text