Hydrocortisone, 98%, Thermo Scientific Chemicals
Hydrocortisone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrocortisone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16292.06
also known as A16292-06
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
Add to cart
Hydrocortisone, 98%, Thermo Scientific Chemicals
Catalog numberA16292.06
Price (USD)/ Each
93.65
Online exclusive
104.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS50-23-7
IUPAC Name(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular FormulaC21H30O5
InChI KeyJYGXADMDTFJGBT-VWUMJDOOSA-N
SMILESC[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
CommentMaterial sourced in the U.S. and in other countries
Appearance (Color)White
Assay (HPLC)≥97.5%
Optical Rotation153.0 ± 3.5° (C=1 in dioxane, non-U.S. sourced material)
View more
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cortisol, steroid hormone is released in response to stress and a low level of blood glucocorticoids. Its primary functions is to increase blood sugar through gluconeogenesis, suppress the immune system, and aid in fat, protein and carbohydrate metabolism.

Solubility
It is soluble in DMSO and ethanol.

Notes
Protect against light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S Spireas .; S Sadu, R Grover. In vitro release evaluation of hydrocortisone liquisolid tablets. Journal of Pharmaceutical Sciences. 1998, 87, (7), 867-872.
  2. P Dandona.; K Thusu, R Hafeez.; E Abdel-Rahman. Effect of hydrocortisone on oxygen free radical generation by mononuclear cells. Metabolism. 1998, 47, (7), 788-791.