Thermo Scientific Chemicals

Tetramethylurea, 99%, Thermo Scientific Chemicals

Catalog number: A16295.30
250 g, Each
Thermo Scientific Chemicals

Tetramethylurea, 99%, Thermo Scientific Chemicals

Catalog number: A16295.30
250 g, Each
Quantity
Catalog number: A16295.30
also known as A16295-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
632-22-4
IUPAC Name
1,1,3,3-tetramethylurea
Molecular Formula
C5H12N2O
InChI Key
AVQQQNCBBIEMEU-UHFFFAOYSA-N
SMILES
CN(C)C(=O)N(C)C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.4500-1.4530 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

Tetramethylurea is used as a solvent in dyestuff industries, in condensation reaction and intermediates in surfactant. It is utilized for base catalyzed isomerization and alkylation hydrocyanation due to its low permittivity. It reacts with oxalyl chloride to prepare tetramethyl chloroformamidinium chloride, which is used for the conversion of carboxylic acids and dialkyl phosphates to anhydrides and pyrophosphates respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylurea is used as a solvent in dyestuff industries, in condensation reaction and intermediates in surfactant. It is utilized for base catalyzed isomerization and alkylation hydrocyanation due to its low permittivity. It reacts with oxalyl chloride to prepare tetramethyl chloroformamidinium chloride, which is used for the conversion of carboxylic acids and dialkyl phosphates to anhydrides and pyrophosphates respectively.

Solubility
Miscible with water.Miscible with water, petroleum ether and common solvents.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text