Zinc sulfide, 98%, Thermo Scientific Chemicals
Zinc sulfide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc sulfide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
5000 g
Catalog number A16296.0B
also known as A16296-0B
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
Add to cart
Zinc sulfide, 98%, Thermo Scientific Chemicals
Catalog numberA16296.0B
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-98-3
IUPAC Namezinc(2+) sulfanediide
Molecular FormulaSZn
InChI KeyDRDVZXDWVBGGMH-UHFFFAOYSA-N
SMILES[S--].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Complexometric Titration≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
FormPowder

Zinc Sulfide is used as a friction material to carry sulfur, which hardens rotor surfaces and thereby reduces wear. Also in printing inks, UV-hardened systems, powder coatings, adhesives, insulating & sealing compounds, thermoplastic pigmentation, thermosets, flame resistant plastics, glass-fiber reinforced plastics, pigment concentrates, elastomers,. textile fibers, paper, mastics, lubricants, electroluminescent lamps, infrared windows, domes and optical elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc Sulfide is used as a friction material to carry sulfur, which hardens rotor surfaces and thereby reduces wear. Also in printing inks, UV-hardened systems, powder coatings, adhesives, insulating & sealing compounds, thermoplastic pigmentation, thermosets, flame resistant plastics, glass-fiber reinforced plastics, pigment concentrates, elastomers,. textile fibers, paper, mastics, lubricants, electroluminescent lamps, infrared windows, domes and optical elements.

Solubility
It is insoluble in water, soluble in acids.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air and moisture sensitive. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. R Vacassy.; SM Scholz.; J Dutta. Synthesis of controlled spherical zinc sulfide particles by precipitation from homogeneous solutions. Journal of the American Ceramic Society. 1998, 81, (10), 2699-2705.
  2. B Ludolph.; MA Malik. Novel single molecule precursor routes for the direct synthesis of highly monodispersed quantum dots of cadmium or zinc sulfide or selenide. Chem. Commun. 1998, 81, (17), 1849-1850.