Thermo Scientific Chemicals

Tetramethylcyclobutane-1,3-dione, 99%, Thermo Scientific Chemicals

Catalog number: A16297.09
10 g, Each
Thermo Scientific Chemicals

Tetramethylcyclobutane-1,3-dione, 99%, Thermo Scientific Chemicals

Catalog number: A16297.09
10 g, Each
Quantity
Catalog number: A16297.09
also known as A16297-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
933-52-8
IUPAC Name
tetramethylcyclobutane-1,3-dione
Molecular Formula
C8H12O2
InChI Key
RGCDVHNITQEYPO-UHFFFAOYSA-N
SMILES
CC1(C)C(=O)C(C)(C)C1=O
Appearance (Color)
White to cream
Assay (GC)
≥98.5%
Melting Point (clear melt)
110.0-117.0?C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

Solubility
It is soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text