Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.
Solubility
It is soluble in chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
RUO – Research Use Only
P Brint.; K Wittel.; P Hochmann.; WS Felps. Molecular Rydberg transitions. 3. A linear combination of Rydberg orbitals (LCRO) model for the two-chromophoric system 2,2,4,4-tetramethylcyclobutane-1,3-dione (TMCBD). J. Am. Chem. Soc. 1976, 98,(25), 7980-7989.
FO Nicolaisen.; OF Nielsen.; M Vala. Vibrational spectra of tetramethylcyclobutane-1, 3-dione. J. Am. Chem. Soc. 1976, 98,(25), 349-359.