Tetramethylcyclobutane-1,3-dione, 99%, Thermo Scientific Chemicals
Tetramethylcyclobutane-1,3-dione, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylcyclobutane-1,3-dione, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number A16297.30
also known as A16297-30
Price (USD)/ Each
594.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
594.00
Add to cart
Tetramethylcyclobutane-1,3-dione, 99%, Thermo Scientific Chemicals
Catalog numberA16297.30
Price (USD)/ Each
594.00
-
Add to cart
Chemical Identifiers
CAS933-52-8
IUPAC Nametetramethylcyclobutane-1,3-dione
Molecular FormulaC8H12O2
InChI KeyRGCDVHNITQEYPO-UHFFFAOYSA-N
SMILESCC1(C)C(=O)C(C)(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥98.5%
Melting Point (clear melt)110.0-117.0?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

Solubility
It is soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
RUO – Research Use Only

General References:

  1. P Brint.; K Wittel.; P Hochmann.; WS Felps. Molecular Rydberg transitions. 3. A linear combination of Rydberg orbitals (LCRO) model for the two-chromophoric system 2,2,4,4-tetramethylcyclobutane-1,3-dione (TMCBD). J. Am. Chem. Soc. 1976, 98,(25), 7980-7989.
  2. FO Nicolaisen.; OF Nielsen.; M Vala. Vibrational spectra of tetramethylcyclobutane-1, 3-dione. J. Am. Chem. Soc. 1976, 98,(25), 349-359.