Thermo Scientific Chemicals

3,4-Dichlorophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A16298.14
25 g, Each
Thermo Scientific Chemicals

3,4-Dichlorophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A16298.14
25 g, Each
Quantity
Catalog number: A16298.14
also known as A16298-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-36-3
IUPAC Name
1,2-dichloro-4-isocyanatobenzene
Molecular Formula
C7H3Cl2NO
InChI Key
MFUVCHZWGSJKEQ-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1Cl)N=C=O
Form
Crystals or fused solid
Appearance (Color)
White to pale cream
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
40-44?C

Description

Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C 'labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C ′″labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, amines, alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text