sodium decyl sulphate is used as a superior dispersing, wetting and emulsifying agent suitable for use in the production of gypsum boards, fire-fighting foams, carpet and upholstery cleaners, special industrial and all-purpose cleaners, oil fields and in the latex manufacture. It is also been used in it to be used in ion pair chromatography and icellar electrokinetic capillary chromatography. other applications include HI&I cleaning, emulsion polymerization, Fire-fighting, Industrial auxiliaries, Construction chemicals, Oil fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
sodium decyl sulphate is used as a superior dispersing, wetting and emulsifying agent suitable for use in the production of gypsum boards, fire-fighting foams, carpet and upholstery cleaners, special industrial and all-purpose cleaners, oil fields and in the latex manufacture. It is also been used in it to be used in ion pair chromatography and icellar electrokinetic capillary chromatography. other applications include HI&I cleaning, emulsion polymerization, Fire-fighting, Industrial auxiliaries, Construction chemicals, Oil fields.
Solubility
It is soluble in H20, poorly soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic, Keepp away from acids Strong oxidizing agents.
RUO – Research Use Only
General References:
- HG Schild.; DA Tirrell.; Langmuir. Interaction of poly (N-isopropylacrylamide) with sodium n-alkyl sulfates in aqueous solution. Langmuir. 1991, 7,(4), 665-671.
- Z Király.; I Dekány. A thermometric titration study on the micelle formation of sodium decyl sulfate in water. Journal of Colloid and Interface Science. 2001, 242,(1), 214-219.