Thermo Scientific Chemicals

(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A16301.36
500 g, Each
Thermo Scientific Chemicals

(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A16301.36
500 g, Each
Quantity
Catalog number: A16301.36
also known as A16301-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1779-58-4
IUPAC Name
(2-methoxy-2-oxoethyl)triphenylphosphanium bromide
Molecular Formula
C21H20BrO2P
InChI Key
VCWBQLMDSMSVRL-UHFFFAOYSA-M
SMILES
[Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Appearance (Color)
White
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%

Description

(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text