Thermo Scientific Chemicals

3,4,5-Trihydroxybenzoic acid monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A16303.0B
1000 g, Each
Thermo Scientific Chemicals

3,4,5-Trihydroxybenzoic acid monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A16303.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16303.0B
also known as A16303-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5995-86-8
IUPAC Name
3,4,5-trihydroxybenzoate
Molecular Formula
C7H5O5
InChI Key
LNTHITQWFMADLM-UHFFFAOYSA-M
SMILES
OC1=CC(=CC(O)=C1O)C([O-])=O
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (on dried substance)
Water Content (Karl Fischer Titration)
≤12.0%
Form
Powder

Description

This chemical is used for the determination of cerium, titanium, phosphate and nitrite with the spectrophotometry. It's also used in the manufacture of thiazine dyes, oxazine dyes, drugs, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This chemical is used for the determination of cerium, titanium, phosphate and nitrite with the spectrophotometry. It′s also used in the manufacture of thiazine dyes, oxazine dyes, drugs, etc.

Solubility
It is soluble in alcohol, ether, glycerol, acetone negligible in benzene, chloroform, petroleum ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Light sensitive. Keep away from Strong oxidizing agents, Strong bases, Acid chlorides, Acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text