Thermo Scientific Chemicals

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A16317.14
25 g, Each
Thermo Scientific Chemicals

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A16317.14
25 g, Each
Quantity
Catalog number: A16317.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149022-15-1
IUPAC Name
2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
Molecular Formula
C8H10ClN3S
InChI Key
OEZPVSPULCMUQB-UHFFFAOYSA-N
SMILES
Cl.CN1C(SC2=CC=CC=C12)=NN
Form
Powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to cream or pale pink
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text