2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.
Solubility
Miscible with carbon tetrachloride, alcohol, chloroform, ether, oils, glacial acetic acid and sodium hydroxide solution. Slightly miscible with water and petroleum ether.
Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
Selvaraj, M.; Selvamani, A.; Alagarasi, A.; Gurulakshmi, M.; Krishnan, P. S.; Suthagar, K.; Shanthi, K. Hydrodeoxygenation of 2-Methoxyphenol Using NiO and NiO-MoO3/Mesoporous Aluminosilicate Catalysts: Effect of MoO3 and aluminum on Activity and Selectivity. Sci. Adv. Mater. 2015, 7 (4), 714-728.
Leiva, K.; Sepulveda, C.; García, R.; Laurenti, D.; Vrinat, M.; Geantet, C.; Escalona, N. Kinetic study of the conversion of 2-methoxyphenol over supported Re catalysts: Sulfide and oxide state. Appl. Catal., A 2015, 505, 302-308.