Thermo Scientific Chemicals

Cyclohexylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: A16320.36
500 g, Each
Thermo Scientific Chemicals

Cyclohexylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: A16320.36
500 g, Each
Quantity
Catalog number: A16320.36
also known as A16320-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
827-52-1
IUPAC Name
cyclohexylbenzene
Molecular Formula
C12H16
InChI Key
IGARGHRYKHJQSM-UHFFFAOYSA-N
SMILES
C1CCC(CC1)C1=CC=CC=C1
Assay (GC)
≥97.0%
Appearance (Color)
Clear colorless
Refractive Index
1.5225-1.5275 @ 20?C
Form
Liquid

Description

Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text