Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals
Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16330.0B
also known as A16330-0B
Price (USD)/ Each
464.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
547.00 
Save 82.35 (15%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
464.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
547.00 
Save 82.35 (15%)
Add to cart
Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals
Catalog numberA16330.0B
Price (USD)/ Each
464.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
547.00 
Save 82.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7783-35-9
IUPAC Namemercury(2+) sulfate
Molecular FormulaHgO4S
InChI KeyDOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES[Hg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or Crystalline powder
Trace Metal(Iron, Fe) : ≤0.005%
Appearance (Color)White
Assay (unspecified)>98.0%
Chloride content≤0.003%
It is used in medicine as a catalyst and in extracting gold and silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine as a catalyst and in extracting gold and silver.

Solubility
It is soluble in hot H2SO4, NaCl solution. insoluble in alcohol, acetone, ammonia

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Light sensitive. Moisture sensitive. Air sensitive. Keep away from Strong oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. A Shaabani.; E Soleimani.; HR Khavasi.; RD Hoffmann. An isocyanide-based three-component reaction: synthesis of fully substituted N-alkyl-2-triphenylphosphoranylidene glutarimides. J. Chem. Educ. 2007, 84, (8), 1273.
  2. BJ Dutka.; N Nyholm.; J Petersen. Comparison of several microbiological toxicity screening tests. Water Research. 1983, 17, (10), 1363-1368.