Thermo Scientific Chemicals

Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals

Catalog number: A16330.30
250 g, Each
Thermo Scientific Chemicals

Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals

Catalog number: A16330.30
250 g, Each
Quantity
Catalog number: A16330.30
also known as A16330-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-35-9
IUPAC Name
mercury(2+) sulfate
Molecular Formula
HgO4S
InChI Key
DOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES
[Hg++].[O-]S([O-])(=O)=O
Appearance (Color)
White
Form
Crystals or Crystalline powder
Trace Metal
(Iron, Fe) : ≤0.005%
Assay (unspecified)
>98.0%
Chloride content
≤0.003%

Description

It is used in medicine as a catalyst and in extracting gold and silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine as a catalyst and in extracting gold and silver.

Solubility
It is soluble in hot H2SO4, NaCl solution. insoluble in alcohol, acetone, ammonia

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Light sensitive. Moisture sensitive. Air sensitive. Keep away from Strong oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text